Skip to Content
Merck
All Photos(1)

Key Documents

CLS3414

Corning® Transwell® 6 well plates

3.0 μm pore size, Tissue Culture (TC)-treated surface round wells, clear bottom, clear wells, flat bottom wells, clear polystyrene (plate or dish), polycarbonate membrane sterile, pack of 6, case of 24

Synonym(s):

cell culture inserts, corning plates, tissue culture plate, tissue culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
€277.00

€277.00


Estimated to ship on31 March 2025



Select a Size

Change View
24 EA
€277.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
clear bottom
clear polystyrene (plate or dish)
clear wells
flat bottom wells
polycarbonate membrane
round wells
pore size:
3.0 μm
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface

€277.00


Estimated to ship on31 March 2025


Product Name

Corning® Transwell® polycarbonate membrane cell culture inserts, 24 mm Transwell with 3.0 μm pore polycarbonate membrane insert, TC-treated, w/lid, sterile, 24/cs

material

clear bottom
clear polystyrene (plate or dish)
clear wells
flat bottom wells
polycarbonate membrane
round wells

sterility

sterile

feature

lid
skirt

packaging

case of 24
pack of 6

manufacturer/tradename

Corning 3414

technique(s)

cell culture | mammalian: suitable

insert working volume

1.5 mL

membrane diam.

24 mm

size

6 wells

surface area

4.67 cm2 , cell growth area

well working volume

2.6 mL

color

translucent

pore size

3.0 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • 10μm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Packaged 6 inserts in a 6 well plate, 4 plates per case
  • Membrane must be stained for cell visibility
  • Sterilized by gamma radiation
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines. They feature a 10 μm thin transparent polycarbonate membrane available in a variety of pore sizes. Transwell are treated for optimal cell attachment. Cells must be stained for cell visibility. The PC membrane is compatible with most organic fixatives and stains.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service