Skip to Content
Merck
All Photos(1)

Key Documents

CLS3335

Corning® CellBIND® Multiple Well Plate

size 6 wells, flat clear bottom, sterile, lid

Synonym(s):

6 well culture plate, cell culture multiwell plates, cell culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
€33.30
50 EA
€238.00

€33.30


Estimated to ship on01 April 2025



Select a Size

Change View
5 EA
€33.30
50 EA
€238.00

About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NB.13
material:
clear polystyrene plate
flat bottom wells clear
flat clear bottom
polystyrene
size:
6 wells
surface area:
9.5 cm2 , cell growth area
well volume:
16.8 mL
well working volume:
1.9-2.9 mL
sterility:
sterile
sterile
binding type:
CellBIND® surface
feature:
lid
lid
skirt
plate format: 6 well standard

€33.30


Estimated to ship on01 April 2025


material

clear polystyrene plate
flat bottom wells clear
flat clear bottom
polystyrene

sterility

sterile
sterile

feature

lid
lid
skirt
plate format: 6 well standard

packaging

pack of 5 ea
case of 50 ea

manufacturer/tradename

Corning 3335

size

6 wells

surface area

9.5 cm2 , cell growth area

well volume

16.8 mL

well working volume

1.9-2.9 mL

suitability

suitable for (cell culture involving anchorage-dependant cell lines)

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® CellBIND® Cell Culture Multiwell Plates

  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jéssica Borghesi et al.
BMC veterinary research, 16(1), 353-353 (2020-09-26)
Tumours in mammary glands represent the most common neoplasia in bitches, as in humans. This high incidence results in part from the stimulation of sex hormones on these glands. Among mammary tumours, inflammatory carcinoma is the most aggressive, presenting a
Min Shen et al.
International immunopharmacology, 85, 106637-106637 (2020-06-09)
More and more evidence showed that autophagy is an inflammation-related defense mechanism against a variety of diseases including liver fibrosis. However, the essential mechanisms remain poorly understood. In this study, we sought to elucidate the impact of Oroxylin A on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service