Key Documents
CLS3081200CO
Pyrex® large knob top desiccators
replacement cover for 5.8L large knob top desiccator, 1/cs
Synonym(s):
desiccator covers, desiccator lids
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3081-200CO
suitability
suitable for (general analytical work)
Related Categories
General description
Note: This is the desiccator replacement cover for Pyrex desiccator CLS3181200.
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)