Skip to Content
Merck
All Photos(2)

Key Documents

CLS139532SS

Corning® Silicone Septa

for use with GL32 Open Top PBT Screw Cap

Synonym(s):

Corning tube cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103800
Pricing and availability is not currently available.

material

natural

sterility

non-sterile

packaging

pack of 10

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Corning silicone septa fit into open top PBT caps (CLS139532HTSC) with GL32 threads to allow for aseptic reagent delivery or culture sampling using a syringe and needle. They can be autoclaved at 12 deg. C or dry heat sterilized at up to 180 deg. C.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service