CLLS1044
MCF10A CELLS GSK3B (-/-)
human female mammary glands (Source Disease: Fibrocystic Disease)
Select a Size
About This Item
Recommended Products
Product Name
MCF10A CELLS GSK3B (-/-),
biological source
human female mammary glands (Source Disease: Fibrocystic Disease)
OMIM accession no.
storage temp.
−196°C
Gene Information
human ... GSK3B(2932)
General description
Application
Features and Benefits
Quality
Disclaimer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service