Skip to Content
Merck
All Photos(1)

Documents

NCI-H69/LX4

96042329, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H69/LX4, 96042329

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian small cell lung carcinoma, drug-resistant

Cell Line Description

NCI-H69/LX4 has been established by exposure of the parent line, H69 (Sigma Catalogue number. 91091802) to doxorubicin, also known as adriamycin. The line hyperexpresses P-glycoprotein and demonstrates a multidrug resistant drug accumulation deficit. It is recommended to culture the cells without drug after resuscitation until the first passage.

Application

Analysis of acquired drug-resistance, study of small cell lung cancer

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 12
D16S539: 11
D5S818: 11,13
D7S820: 9
THO1: 8,9
TPOX: 10
vWA: 16,17

Culture Medium

RPMI + 2mM Glutamine + 0.4μg/ml doxorubicine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) by repeated pipetting of aggregates or scraping; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service