HR5
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human cervix
packaging
tube of 5 μg 95091409-DNA-5UG
pkg of vial of cells 95091409-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human cervix carcinoma, HeLa derivative, transformed
Cell Line Description
HR5 cells were obtained after transfection of HeLa cells with plasmids coding for a reverse tetracycline-controlled transactivator encoding a nuclear localisation sequence (rt TA-nls) and transferring G418 resistance. This transactivator (a fusion between the Tet repressor and the activating domain of the herpes simplex virus VP16 transcription activation domain) can stimulate the expression of a reporter gene up to 10E5-fold. HR5 is the parent cell line for HR5-CL11 (Sigma Catalogue number.95091411).
Application
Gene expression and activation studies
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 to 1:20 i.e. seeding at 2x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service