Skip to Content
Merck
All Photos(1)

Key Documents

ATHOS

93122325, Porcine thyroid, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

ATHOS, 93122325

biological source

Porcine thyroid

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Thyroglobulin, plasminogen activator (u-PA)

receptors

Thyrotropin, insulin, Insulin Growth Factor (IGF-1), hydrocortisone

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Porcine thyroid epithelial

Cell Line Description

The cell line was spontaneously derived from the porcine thyroid cell line PORTHOS (Sigma Catalogue number. 93122323) after withdrawal of thyroid stimulating hormone (TSH). The cells form follicle-like structures, have a high growth rate, a high glucose consumption and are TSH independent for growth. The expression of plasminogen activators has been reported and found to be higher in ATHOS cells compared to PORTHOS cells.

Application

Mechanism of thyroid follicle reconstruction

Culture Medium

Click:RPMI (1:1) + 10μg/ml Insulin + 10nM Hydrocortisone + 5μg/ml Transferrin + 10ng/ml glycyl-L-histydyl-L-lysine acetate + 10ng/ml somatostatin + 2mM Glutamine + 0.1% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 5x100,000 cells/cm2 using 0.05% trypsin/0.025% EDTA in Puck F saline; 2.5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service