Skip to Content
Merck
All Photos(1)

Documents

DBTRG.05MG

93061119, human brain, Glial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DBTRG.05MG, 93061119

biological source

human brain

growth mode

Adherent

karyotype

2n = 87-91, hypotetraploid

morphology

Glial

products

Not specified

receptors

Epidermal growth factor (EGF) R

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human glioblastoma

Cell Line Description

Derived from an adult female with glioblastoma multiforme treated with local brain irradiation and multi drug chemotherapy. By flow cytometry these cells were almost 100% positive for MHC Class I.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 9
D16S539: 12
D5S818: 12,13
D7S820: 11
THO1: 8
TPOX: 8
vWA: 15,16

Culture Medium

RPMI 1640 + 2mM Glutamine + 1% HT + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service