Skip to Content
Merck
All Photos(1)

Key Documents

HT2 Clone A5E

92021401, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

HT2 Clone A5E, 92021401, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 92021401, mouse blood, Lymphoblast

biological source

mouse blood

packaging

tube of 5 μg 92021401-DNA-5UG
pkg of vial of cells 92021401-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Interleukin 2 (IL-2), Interleukin 4 (IL-4)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse helper T cell

Cell Line Description

Derived in 1979 from BALB/c spleen cells activated by sheep erythrocytes in the presence of IL-2, the A5E clone was selected to provide a homogenous population that is highly sensitive to IL-2. This cell line expresses Thy-1 and Lyt markers in addition to high affinity IL-2 receptors. Substantial proliferation is seen in the presence of IL-4 (GM-CSF).

Application

IL-2 assays

Culture Medium

RPMI 1640 + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 100-200 IU/ml IL-2 + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. When starting from a frozen ampoule, add thawed cells to a conical based centrifuge tube e.g. 15ml tube. Slowly add 4 ml of culture medium to the tube. Take a sample of the cell suspension, e.g

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service