Skip to Content
Merck
All Photos(1)

Documents

SW 948

91030714, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SW 948, 91030714

biological source

human colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

Derived from a grade III adenocarcinoma of the colon of an 81 year old Caucasian female. The cells synthesise relatively large amounts of CEA (2,000ng/1,000,000 cells in 21 days).

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 10,11
D16S539: 11,12
D5S818: 11
D7S820: 9,11
THO1: 6,9.3
TPOX: 8,11
vWA: 16,18

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; No CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service