Skip to Content
Merck
All Photos(1)

Key Documents

RR1022

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

rat unknown/unspecified

packaging

tube of 5 μg 90102526-DNA-5UG
pkg of vial of cells 90102526-1VL

growth mode

Adherent

karyotype

2n = 42

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Amsterdam Schmidt-Ruppin sarcoma

Cell Line Description

Derived from a Rous sarcoma virus (Schmidt-Ruppin strain) induced tumour in an inbred Amsterdam rat. The cells are highly tumorigenic in new-born rats and also chickens. There is some evidence that the cell line is latently infected with the virus.

Application

Tumourigenicity and virus studies: Rous sarcoma virus

Culture Medium

Medium 199 (HBSS) + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service