Skip to Content
Merck
All Photos(1)

Documents

YBM 6.1.10

89040601, Reactivity: Anti-MAC-1, mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

ImmunoDon

DA rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-MAC-1, mouse

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Y3Ag1.2.3 x Rat DA spleen

Cell Line Description

The antibody is directed against the mouse MAC-1 antigen, the murine equivalent of CD11b.

Immunogen

CBA mouse bone marrow

Culture Medium

IDMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells may adhere to the plastic flask - may require trypsin or shaking to remove them

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service