Skip to Content
Merck
All Photos(1)

Key Documents

COS-1 Cell Line

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

African green monkey kidney

packaging

tube of 5 μg 88031701-DNA-5UG
pkg of vial of cells 88031701-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Monkey African green kidney, SV40 transformed

Cell Line Description

Derived from CV-1 simian cells transformed by origin-defective mutant of SV40 which codes for wildtype T Ag. Possible host for propagation of pure populations of recombinant SV40 virus. The cells should be handled under laboratory containment level 2.

Application

COS-1 cell line has been used to test the toxicity of various concentrations of iron oxide nanoparticles (for use in medical diagnostics).[1] It has also been used to study the interaction of nanoparticles with their cellular targets.[2]
Virus replication studies: SV40

Culture Medium

DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).

Subculture Routine

Split semi-confluent cultures 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C. In order to prevent syncitia formation, do not allow cells become confluent.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–7 of 7 Questions  
  1. Are my Cell Line cells mycoplasma tested?

    1 answer
    1. ECACC routinely tests all manufactured cell banks for mycoplasma.

      Helpful?

  2. Why is Vapor Phase Liquid Nitrogen preferred for storage of Cell Line cells?

    1 answer
    1. If ampules are immersed into liquid phase of liquid nitrogen, it increases the risk of the liquid seeping into the vial. This could lead to problems of cross-contamination and increased risk of the ampule exploding when thawed.

      Helpful?

  3. What medium should I use when I thaw the Cell Line cells?

    1 answer
    1. The medium for each cell line is listed on the product page. It is specific to each cell line. If not visible on the Sigma-Aldrich website product page, the HPA website also contains the same information (www.hpacultures.org.uk). The product number for the cell line is the same on either site.

      Helpful?

  4. What passage are my Cell Line cells?

    1 answer
    1. If the passage of the cells is known, it is listed on the product page at the HPA website (www.hpacultures.org.uk).

      Helpful?

  5. Are the Cell Line cells tested for viral pathogens?

    1 answer
    1. HPA Cultures does not perform any viral testing on the cell lines.

      Helpful?

  6. How do I handle Cell Line frozen cells upon arrival?

    1 answer
    1. Upon receipt, frozen ampules should be transferred directly to vapor phase liquid nitrogen without delay (-135°C) or liquid phase liquid nitrogen if vapor phase is not available. DO NOT use a -80°C freezer as an alternative; this will result in loss of viability.

      Helpful?

  7. How many Cell Line cells are in the vial?

    1 answer
    1. Each vial of cells contains 2-3 x 106 cells in 1 ml of freezing media. This is in a 1.8 mL ampule.  For exceptionally large cells, counts may decrease. Suspension cells, generally smaller cells, may contain as many as 4-5 x 106 cells/vial to assure optimal viability upon thaw.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service