Skip to Content
Merck
All Photos(1)

Documents

P1.HTR.TK-

87121103, mouse breast, Mastocytoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P1.HTR.TK-, 87121103

biological source

mouse breast

growth mode

Suspension

karyotype

Not specified

morphology

Mastocytoma

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse DBA/2 mastocytoma

Cell Line Description

Derived from the mouse mastocytoma P815 after serial cycles of transfection with a plasmid carrying a TK gene. BUdR is necessary in the growth media to avoid revertants.

Application

Genetic, transfection and tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 100ug/ml Bromo-deoxy Uridine (BUdR) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 8% CO2; 37°C. Media change twice a week.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service