Skip to Content
Merck
All Photos(1)

Key Documents

Y79

86093003, human eye, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

Y79, 86093003

biological source

human eye

growth mode

Suspension

karyotype

2n = 46, hypertriploid

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian retinoblastoma

Cell Line Description

Established from a primary tumour of the right eye of a 2.5 year old female Caucasian. The cells grow in clusters and exhibit reverse transcriptase activity.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 4-9x100,000 cells/ml; 5% CO2; 37°C.When in culture, stand flask up until cells show growth in large clumps. After approximately 3 days the flask can be laid down.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service