Skip to Content
Merck
All Photos(1)

Key Documents

QIMR-WIL

86030601, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

QIMR-WIL, 86030601

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Epstein Barr Virus (EBV)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human myeloblastic leukaemia lymphoblast

Cell Line Description

Established from peripheral lymphocytes of a patient with myeloblastic leukaemia at the Queensland Institute for Medical Research. It produces EBV and has been used in co-cultivation lymphocyte transformation studies. Non-uniform lymphoblast.

Application

Transformation studies

Culture Medium

DMEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures at 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service