Skip to Content
Merck
All Photos(1)

Documents

LIM2405 cell line human

12062003, human colon, epithelial

Synonym(s):

LIM 2405, LIM-2405

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LIM2405 cell line human, 12062003

biological source

human colon

growth mode

adherent

morphology

epithelial

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Cell Line Description

Human adenocarcinoma of the caecum of a male patient. Cell Bank Australia reference number: CBA0165.

Culture Medium

RPMI-1640 (with 2mM L-Glutamine + 25mM HEPES) + 10% FCS, Insulin 0.6 μg/ml, Hydrocortisone 1μg/ml, 1-Thioglycerol 10 μM

Subculture Routine

Optimal split ratio 1:4-1:6 (seeding density 1.6 x 104 cells/cm2). Cells incubated at 37°C with 5% CO2. Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5 min. LIM2405 may take 2-3 days to seed after thawing or trypsinisation.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service