Skip to Content
Merck
All Photos(1)

Key Documents

ZL34

11120713, human lung, Mesenchymal with thin cytoplasmic processes

Synonym(s):

ZL 34, ZL-34

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

ZL34, 11120713

biological source

human lung

growth mode

Adherent

karyotype

Not specified

morphology

Mesenchymal with thin cytoplasmic processes

receptors

Not specified

relevant disease(s)

cancer

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure

Cell Line Description

ZL34 was derived from a fibrous tumour of the pleural cavity in a 53-year old male patient, prior to treatment, with a known history of exposure to asbestos.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 11,13
D13S317: 11,12
D16S539: 11
D5S818: 11
D7S820: 9
THO1: 6
TPOX: 8
vWA: 18,20

Culture Medium

DMEM:F12 (1:1) (D8062) + L-Glutamine (G7513) + 15% FBS / FCS (F2442).

Subculture Routine

Split subconfluent cultures (70-80%) 1:4 to 1:10 using 0.25% trypsin/EDTA; 5% CO2; 37 °C. Suggested seeding density 1-4 x 10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service