Skip to Content
Merck
All Photos(1)

Documents

PNMR 160D

Reactivity: Anti-Neural Crest (Quail) and human neuroendocrine tissue, 01080923

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

ImmunoDon

BALB/c mice

Myeloma

SP2

Reactivity

Anti-Neural Crest (Quail) and human neuroendocrine tissue

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Anti-Quail Neural Crest Hybridoma

Cell Line Description

This antibody may provide a valuable tool for studying the differentiation of embryonic precursor cells. The hybridoma supernatant cross reacts with human neuroendocrine cells within appendix, colon and rectum. This mab could provide novel reagents for understanding neural crest development. The antibody is not reactive with somite cultures. The purified antibody is best used at 30 μg/ml for immunocytochemistry. Hybridoma should produce 1-60 μg/ml.

Immunogen

Quail Neural Crest

Application

Immunocytochemistry, diagnostic tools and probes for cellular and macromolecular investigations

Subculture Routine

Allow hydridomas to become 75% confluent prior to splitting. Seed at 3-9 x 100,000 cells/ml, 5% CO2, 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service