Skip to Content
Merck

Skip To

C9847

Cyclothiazide

AMPA glutamate receptor blocker, powder

Synonym(s):

6-Chloro-3,4-dihydro-3-(2-norbornen-5-yl)-2H-1,2-4-benzothiadiazine-7-sulfonamide 1,1-dioxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 119,00
25 mg
Check Cart for Availability
€ 283,00

About This Item

Empirical Formula (Hill Notation):
C14H16ClN3O4S2
CAS Number:
Molecular Weight:
389.88
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352125
EC Number:
218-859-7
MDL number:
Form:
powder

€ 119,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cyclothiazide,

form

powder

Quality Segment

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

[H][C@@]12CC(C3Nc4cc(Cl)c(cc4S(=O)(=O)N3)S(N)(=O)=O)[C@@]([H])(C1)C=C2

InChI

1S/C14H16ClN3O4S2/c15-10-5-11-13(6-12(10)23(16,19)20)24(21,22)18-14(17-11)9-4-7-1-2-8(9)3-7/h1-2,5-9,14,17-18H,3-4H2,(H2,16,19,20)/t7-,8+,9?,14?/m0/s1

InChI key

BOCUKUHCLICSIY-QJWLJZLASA-N

General description

Cyclothiazide is a benzothiadiazine, which has a similar structure to diazoxide.[1]

Application

Cyclothiazide has been used as a α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) desensitization blocker to study the effects of γ2 on receptor desensitization.[2]

Biochem/physiol Actions

Blocks the rapid desensitization of the AMPA glutamate receptors and markedly prolongs the decay time of the evoked excitatory post-synaptic current.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D9035C4911SML2047
form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

originator

Eli Lilly

originator

Schering Plough

originator

-

originator

-

Gene Information

human ... CA1(759), CA4(762), GRIA1(2890), GRIA2(2891), GRIA3(2892), GRIA4(2893)
rat ... Gria1(50592)

Gene Information

human ... KCNA1(3736), KCNJ11(3767), KCNJ8(3764), KCNMA1(3778)
mouse ... Kcnj1(56379), Kcnj11(16514)
rat ... Gria1(50592), Kcna1(24520), Kcnj1(24521), Kcnj11(83535), Kcnj8(25472)

Gene Information

human ... CA1(759), CA4(762), SLC12A3(6559)

Gene Information

-


Still not finding the right product?

Explore all of our products under Cyclothiazide


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters