Skip to Content
Merck
All Photos(5)

Key Documents

C9616

Sigma-Aldrich

Anti-Cytochrome c antibody produced in sheep

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CYC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

sheep

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15 kDa

species reactivity

rabbit, rat, human, canine

concentration

~0.5 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 20-40 μg/mL using human heart tissue
indirect immunofluorescence: 5-10 μg/mL using human MCF-7 cells
western blot: 0.1-0.2 μg/mL using whole extracts of MCF−7, Jurkat, Rat−1, MDCK cells and extract of rat kidney or rat heart

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYCS(54205)
rat ... Cycs(25309)

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Cytochrome c produced in sheep using purified rabbit cytochrome c conjugated to keyhole limpet hemocyanin (KLH).

Immunogen

rabbit cytochrome c.

Application

Anti-Cytochrome c antibody produced in sheep has been used in:
  • immunocytochemistry
  • immunostaining
  • immunoblotting
  • immunohistochemistry

Biochem/physiol Actions

Cytochrome c controls cellular electron transport and energy metabolism. It is involved in the transfer of electrons between complex III and complex IV in the mitochondrial electron transport chain. It is involved in apoptosis and activates the death protease caspase-3 (CCP32). The presence of Bcl-2 on the organelles inhibits the release of cytochrome c from mitochondria during apoptosis. Along with Apaf-1, and procaspase-9, it forms an essential component of vertebrate ”apoptosome”. Activation of caspase-9 and other capsases directs apoptosis. Serum cytochrome c is a sensitive apoptotic marker in vivo, and increased serum cytochrome c level can serve as a negative prognostic marker.

Target description

Cytochrome c is an electron transport protein released from mitochondria as an early committed event in apoptosis. Cytochrome c and dATP are cofactors for the mammalian apoptosome, which is composed of Apaf-1, Bcl-2, and procaspase 9. When caspase 9 is activated, activation of other caspases follow including the death protease caspase 3. The release of cytochrome c is inhibited by the presence of Bcl-2 on these organelles preventing the initiation of apoptosis. In cells induced by several apoptotic agents (such as UV irradiation, staurosporine, and overexpression of Bax), caspase inhibitors do not prevent cytochrome c release.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Barczyk et al.
International journal of cancer, 116(2), 167-173 (2005-04-01)
Despite significant progress in cancer therapy, the outcome of the treatment is often unfavorable. Better treatment monitoring would not only allow an individual more effective, patient-adjusted therapy, but also it would eliminate some of the side effects. Using a cytochrome
Differential control of growth, apoptotic activity and gene expression in human colon cancer cells by extracts derived from medicinal herbs, Rhazya stricta and Zingiber officinale and their combination
Elkady AI, et al.
World Journal of Gastroenterology, 20(41), 15275-15275 (2014)
R M Kluck et al.
Science (New York, N.Y.), 275(5303), 1132-1136 (1997-02-21)
In a cell-free apoptosis system, mitochondria spontaneously released cytochrome c, which activated DEVD-specific caspases, leading to fodrin cleavage and apoptotic nuclear morphology. Bcl-2 acted in situ on mitochondria to prevent the release of cytochrome c and thus caspase activation. During
Coupling of phosphorylation to electron and hydrogen transfer by a chemi-osmotic type of mechanism.
P MITCHELL
Nature, 191, 144-148 (1961-07-08)
P Li et al.
Cell, 91(4), 479-489 (1997-12-09)
We report here the purification of the third protein factor, Apaf-3, that participates in caspase-3 activation in vitro. Apaf-3 was identified as a member of the caspase family, caspase-9. Caspase-9 and Apaf-1 bind to each other via their respective NH2-terminal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service