Skip to Content
Merck
All Photos(2)

Key Documents

C9400

Sigma-Aldrich

Calcium Ionophore A23187 hemimagnesium salt

Synonym(s):

A23187 hemimagnesium salt, Antibiotic A 23187 hemimagnesium salt, Calcimycin hemimagnesium salt, Calimycin hemimagnesium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€224.00

€224.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€224.00

About This Item

Empirical Formula (Hill Notation):
C29H36Mg0.5N3O6
CAS Number:
Molecular Weight:
534.77
Beilstein:
6765085
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€224.00


Estimated to ship on16 April 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

antibiotic activity spectrum

Gram-positive bacteria
fungi

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

CNc1ccc2oc(C[C@H]3O[C@@]4(CC[C@H]3C)O[C@@H]([C@H](C)C[C@H]4C)[C@H](C)C(=O)c5ccc[nH]5)nc2c1C(=O)O[Mg]OC(=O)c6c(NC)ccc7oc(C[C@H]8O[C@@]9(CC[C@H]8C)O[C@@H]([C@H](C)C[C@H]9C)[C@H](C)C(=O)c%10ccc[nH]%10)nc67

InChI

1S/2C29H37N3O6.Mg/c2*1-15-10-11-29(17(3)13-16(2)27(38-29)18(4)26(33)20-7-6-12-31-20)37-22(15)14-23-32-25-21(36-23)9-8-19(30-5)24(25)28(34)35;/h2*6-9,12,15-18,22,27,30-31H,10-11,13-14H2,1-5H3,(H,34,35);/q;;+2/p-2/t2*15-,16-,17-,18-,22-,27+,29+;/m11./s1

InChI key

XBWKBTZDBYFEMH-UIOMRPQBSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ionophore highly selective for Ca2+. Potentiates responses to NMDA but not quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase. Used to study effects of elevated intracellular calcium on mitochondrial function and intrinsic apoptosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service