Skip to Content
Merck
All Photos(1)

Key Documents

C9359

Sigma-Aldrich

Z-Ala-Ser-Thr-Asp(OMe)-fluoromethyl ketone

≥90% (TLC), solid

Synonym(s):

EMAP II inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33N4O10F
CAS Number:
Molecular Weight:
556.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (TLC)

form

solid

storage temp.

−20°C

SMILES string

COC(=O)C[C@H](NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](C)NC(=O)OCc1ccccc1)[C@@H](C)O)C(=O)CF

InChI

1S/C24H33FN4O10/c1-13(26-24(37)39-12-15-7-5-4-6-8-15)21(34)28-17(11-30)22(35)29-20(14(2)31)23(36)27-16(18(32)10-25)9-19(33)38-3/h4-8,13-14,16-17,20,30-31H,9-12H2,1-3H3,(H,26,37)(H,27,36)(H,28,34)(H,29,35)/t13-,14+,16-,17-,20-/m0/s1

InChI key

LICIOVKJUZZCAM-ACSCVQKFSA-N

Amino Acid Sequence

Z-Ala-Ser-Thr-Asp(OMe)-FMK

Biochem/physiol Actions

Cell-permeable inhibitor of endothelial monocyte-activating polypeptide II (EMAP-II).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U E Knies et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(21), 12322-12327 (1998-10-15)
Endothelial monocyte-activating polypeptide II (EMAP II) is a proinflammatory cytokine and a chemoattractant for monocytes. We show here that, in the mouse embryo, EMAP II mRNA was most abundant at sites of tissue remodeling where many apoptotic cells could be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service