Skip to Content
Merck

C9024

Cyanocorrin−Agarose

saline suspension

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

saline suspension

extent of labeling

0.3-0.5 mg per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

carboxyl

matrix spacer

8 atoms

capacity

1-3 mg/mL binding capacity (nonintrinsic factor (N 1883))

storage temp.

2-8°C

Application

Cyanocorrin-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and specialty resins. Cyanocorrin has been used in studies to determine electron densities of B12 vitamins.

Physical form

Suspension in 0.5 M NaCl containing preservative


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library