Skip to Content
Merck
All Photos(1)

Documents

C8862

SAFC

EX-CELL® CHO DHFR Medium

Animal-component free, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

product line

EX-CELL®

form

liquid

technique(s)

cell culture | mammalian: suitable (suspension)

components

phenol red: no
glucose: yes
NaHCO3: yes
HEPES: yes
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Related Categories

Application

This animal component-free medium (containing no hypoxanthine or thymidine) is formulated to maximize cell growth and recombinant protein production using the Dihydrofolate Reductase (DHFR) gene amplification system in DHFR- Chinese Hamster Ovary (CHO) cells.

Features and Benefits

Developed to meet the needs of biotechnology and vaccine manufacturing, this medium supports rapid initial cell growth and high levels of protein expression in suspension cultures. This medium supports and maintains high cell densities for extended periods, resulting in increased productivity.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MGAT1 adds N-acetylglucosamine to the Man5GlcNAc2 (Man5) structure. Goh et al. reported increased sialylation after restoring MGAT1 function in MGAT1 deficient CHO cells.

In the present study, we have identifi ed species-specifi c housekeeping genes (HKGs) for Chinese Hamster Ovary (CHO) cells using data from microarray gene expression profiling.

A signal peptide is a 5-30 amino acid (aa) peptide present at the N-terminus of secretory proteins.

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service