Skip to Content
Merck
All Photos(1)

Documents

C8427

Sigma-Aldrich

2′,7′-bis(2-Carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester Mixed isomers

Synonym(s):

BCECF-acetoxymethyl

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CCc1cc2c(Oc3cc(OCOC(C)=O)c(CCC(=O)OCOC(C)=O)cc3C24OC(=O)c5cc(ccc45)C(=O)OCOC(C)=O)cc1OCOC(C)=O

InChI

1S/C42H40O21/c1-22(43)52-17-57-34-15-36-32(13-27(34)7-10-38(48)59-19-54-24(3)45)42(31-9-6-29(12-30(31)41(51)63-42)40(50)61-21-56-26(5)47)33-14-28(8-11-39(49)60-20-55-25(4)46)35(16-37(33)62-36)58-18-53-23(2)44/h6,9,12-16H,7-8,10-11,17-21H2,1-5H3

InChI key

NTECHUXHORNEGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Material is an isomeric mixture of lactones, acetates, and AM esters

Other Notes

Membrane-permeable fluorescent probe for cytoplasmic pH.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sen Xu et al.
Analytical chemistry, 83(4), 1480-1483 (2011-01-20)
Detection of viable pathogenic bacteria has widespread application in food safety and human health. Antibody-based methods require a growth step which limits time-to-results performance. In this study, we use a mass-change sensitive cantilever biosensor and a probe, 2',7'-bis-(2-carboxyethyl)-5-(and-6)-carboxyfluorescein acetoxymethyl ester
Wen Liu et al.
American journal of physiology. Renal physiology, 302(1), F205-F215 (2011-10-01)
Epithelial Na(+) channel (ENaC)-mediated Na(+) absorption and BK channel-mediated K(+) secretion in the cortical collecting duct (CCD) are modulated by flow, the latter requiring an increase in intracellular Ca(2+) concentration ([Ca(2+)](i)), microtubule integrity, and exocytic insertion of preformed channels into
Jacek Lubelski et al.
The Journal of biological chemistry, 279(33), 34449-34455 (2004-06-12)
Multidrug resistance (MDR)-type transporters mediate the active extrusion of structurally and functionally dissimilar compounds from the cells, thereby rendering cells resistant to a range of drugs. The ydaG and ydbA genes of Lactococcus lactis encode two ATP-binding cassette half-transporters, which
Clark Undem et al.
PloS one, 7(9), e46303-e46303 (2012-10-03)
Excessive production of endothelin-1 (ET-1), a potent vasoconstrictor, occurs with several forms of pulmonary hypertension. In addition to modulating vasomotor tone, ET-1 can potentiate pulmonary arterial smooth muscle cell (PASMC) growth and migration, both of which contribute to the vascular
Arno Hueber et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 243(9), 942-946 (2005-04-19)
This study was performed to evaluate the safety and efficacy of photodynamic therapy with the carboxyfluorescein ester BCECF-AM as an adjunctive treatment procedure for pterygium surgery to reduce the rate of recurrence. In this nonrandomized prospective clinical trial, 19 eyes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service