This product is typically used as a nutritive component in various microbiological media. This product is not tested for suitability. The researcher would have to determine the appropriate dilution for their specific requirements. However, you may want to review the Corn Steep Liquor product information. 'A viscous concentrate of corn soluble, rich in vitamins, amino acids, minerals and other growth stimulants. Contains approx. 50% (w/w) solids.'. Corn steep liquor medium (see ATCC 1716 formulation) included 50 ml/L of corn steep liquor. Please see the link below to this product page.
https://www.sigmaaldrich.com/product/sigma/c4648
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
500 g | Check Cart for Availability | € 139,00 |
About This Item
NACRES:
NA.56
UNSPSC Code:
12352202
Form:
powder
Biological source:
corn
biological source
corn
Quality Segment
form
powder
nitrogen analysis
7.0-8.5% total
technique(s)
microbiological culture: suitable
Application
Corn steep solids are an inexpensive nutritional supplement for a wide variety of industrial microbiological processes. For example, this material was used in production of cellulase from a fungal endophyte, as an adjunct to bioethanol production.
Corn steep solids are soluble and may be used as a nitrogen source in several medium and cell culture conditions. This can give rise to biomasses that are rich in proteins and poor in acidic polysaccharides
Other Notes
Spray-dried corn solubles.
1 of 1
This Item | |||
|---|---|---|---|
| description spray-dried corn solubles | description - | description NutriSelect® Basic, powder, for yeasts, for bacteria, for molds, pack of 250 g, pkg of 1 kg | description NutriSelect® Plus, powder, for bacteria, pack of 500 g |
| technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable |
| biological source corn | biological source corn | biological source - | biological source - |
| form powder | form suspension | form powder | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level 200 | Quality Level 200 |
| nitrogen analysis 7.0-8.5% total | nitrogen analysis - | nitrogen analysis - | nitrogen analysis - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What proportion do you need to dilute?
1 answer-
Helpful?
-



