Skip to Content
Merck
All Photos(1)

Key Documents

C7870

Sigma-Aldrich

Chalcone 4 hydrate

≥98% (HPLC)

Synonym(s):

(E)-1-(4-Chlorophenyl)-3-(4-hydroxy-3-methoxyphenyl)-2-propen-1-one hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClO3 · xH2O
CAS Number:
Molecular Weight:
288.73 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

O.COc1cc(ccc1O)\C=C\C(=O)c2ccc(Cl)cc2

InChI

1S/C16H13ClO3.H2O/c1-20-16-10-11(3-9-15(16)19)2-8-14(18)12-4-6-13(17)7-5-12;/h2-10,19H,1H3;1H2/b8-2+;

InChI key

PXVQKTVKTBMRIW-VOKCZWNHSA-N

Biochem/physiol Actions

Chalcone 4 is a potent, selective inhibitor of the chemokine CXCL12 via direct binding. It is also a small molecule anti-ligand (or neutraligand) of CXCR4 and CXCR7. The chemokine CXCL12 (stromal cell-derived factor-1) and its receptors play pivotal roles in inflammatory responses, normal vascular and neuronal development, as well as in infectious diseases and cancer. Chalcone 4 binds to and activates the chemokine receptors CXCR4 and CXCR7 and stimulates a rapid receptor-mediated intracellular calcium mobilization and signaling through a Pertussis toxin sensitive Gi protein. Chalcone 4 potently and selectively inhibits CXCL12 binding to CXCR4 and CXCR7, and consequent CXCL12-evoked calcium cellular responses. Chalcone 4 binds to the chemokine CXCL12, but not to its receptors.

Features and Benefits

This compound is featured on the Chemokine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lalita Kulmala et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 37(6), 538-542 (2014-03-14)
The aim of this study was to determine the predictive value of the combination of plasma-soluble fms-like tyrosine kinase 1 (sFlt-1) and uterine artery Doppler for the detection of preeclampsia in women of advanced age at 16-18 weeks of gestation
Anna Chi Shan Kam et al.
Journal of medical screening, 21(2), 71-75 (2014-03-14)
To establish the reliability and validity of an automated hearing screening test system for preschoolers and to investigate the risk factors for hearing loss. The study used a cross-sectional design in a comparative study of subjects. The automated hearing screening
Karen Zwaenepoel et al.
Histopathology, 65(4), 539-548 (2014-03-14)
Testing for ALK rearrangements in advanced, non-squamous non-small-cell lung cancers that are wild-type for activating EGFR mutation has become standard care. Fluorescence in-situ hybridization is considered the gold standard for this evaluation. Pre-screening with immunohistochemistry has been suggested, to reduce
Susumu Hijioka et al.
Pancreas, 43(3), 367-372 (2014-03-14)
This study investigated whether a risk assessment nomogram can predict the malignant potential of intraductal papillary mucinous neoplasms (IPMNs) and provide valuable information for the follow-up and counseling strategies of such patients. We studied 126 of 589 patients with IPMN
Alexander Kaltenborn et al.
PloS one, 9(3), e91560-e91560 (2014-03-14)
This study introduces and validates the Hip Lag Sign, a new clinical parameter to determine hip abductor damage, which appears to be one major cause for greater trochanteric pain syndrome. 26 patients who underwent standardized MRI-examination were prospectively enrolledbetween October

Articles

We offer many products related to chemokine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service