Skip to Content
Merck
All Photos(1)

Key Documents

C7795

Sigma-Aldrich

Chloramphenicol

γ-irradiated

Synonym(s):

D-(−)-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, D-(−)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]acetamide, Chloromycetin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCONHCH(CH2OH)CH(OH)C6H4NO2
CAS Number:
Molecular Weight:
323.13
Beilstein:
2225532
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.31

Quality Level

sterility

γ-irradiated

form

powder

pKa 

~5.5

mp

149-153 °C (lit.)

solubility

absolute ethanol: 5-20 mg/mL (as a stock solution)
methanol: soluble (as a stock solution)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)/t8-,9-/m1/s1

InChI key

WIIZWVCIJKGZOK-RKDXNWHRSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: phenicole

Application

Chloramphenicol is a synthetic antibiotic, isolated from strains of Streptomyces venezuelae. It is often used for bacterial selection in molecular biology applications at 10-20 μg/mL and as a selection agent for transformed cells containing chloramphenicol reistance genes.

Biochem/physiol Actions

Mode of Action: Chloramphenicol inhibits bacterial protein synthesis by blocking the peptidyl transferase step by binding to the 50S ribosomal subunit and preventing attachment of aminoacyl tRNA to the ribosome. It also inhibits mitochondrial and chloroplast protein synthesis and ribosomal formation of (p)ppGpp, de-pressing rRNA transcription.

Mode of Resistance: Use of chloramphenicol acetyltransferase will acetylate the product and inactivate it.

Antimicrobial Spectrum: This is a broad spectrum antibiotic against gram-positive and gram-negative bacteria, and is used mainly for ophthalmic and veterinary purposes.

Caution

Stock solutions should be stored at 2-8°C and are stable at 37°C for 5 days. Aqueous solutions are neutral and stable over a wide pH range, with 50% hydrolysis occurring after 290 days. Use of a borax buffered solution reduces this number to 14%. Solutions should be protected from light as photochemical decomposition results in a yellowing of the solution. Heating aqueous solutions at 115°C for 30 minutes results in a 10% loss of chloramphenicol.

Preparation Note

Stock solutions can be prepared directly in the vial at any concentration in the recommended range. A solution at 50 mg/mL in ethanol yields a clear, very faint, yellow solution. Degradation of chloramphenicol in aqueous solution is catalyzed by general acids and bases. This rate of degradation is independent of the ionic strength and pH.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Light sensitive.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uwe Richter et al.
Current biology : CB, 23(6), 535-541 (2013-03-05)
Proliferating cells require coordinated gene expression between the nucleus and mitochondria in order to divide, ensuring sufficient organelle number in daughter cells [1]. However, the machinery and mechanisms whereby proliferating cells monitor mitochondria and coordinate organelle biosynthesis remain poorly understood.
Chloramphenicol eye drops: a dangerous drug?
J Diamond et al.
The Practitioner, 239(1555), 608-611 (1995-10-01)
David H Kwan et al.
Journal of the American Chemical Society, 137(17), 5695-5705 (2015-04-15)
Blood transfusions are critically important in many medical procedures, but the presence of antigens on red blood cells (RBCs, erythrocytes) means that careful blood-typing must be carried out prior to transfusion to avoid adverse and sometimes fatal reactions following transfusion.
David G Kirk et al.
Applied and environmental microbiology, 80(16), 5141-5150 (2014-06-15)
Clostridium botulinum produces heat-resistant endospores that may germinate and outgrow into neurotoxic cultures in foods. Sporulation is regulated by the transcription factor Spo0A and the alternative sigma factors SigF, SigE, SigG, and SigK in most spore formers studied to date.
J N de Almeida Júnior et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 20(8), 784-790 (2013-12-21)
Trichosporon spp. have recently emerged as significant human pathogens. Identification of these species is important, both for epidemiological purposes and for therapeutic management, but conventional identification based on biochemical traits is hindered by the lack of updates to the species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service