Skip to Content
Merck
All Photos(1)

Key Documents

C7278

Sigma-Aldrich

Caseinoglycopeptide from bovine casein

essentially salt-free, lyophilized powder

Synonym(s):

Bovine Casein Glycopeptide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€733.00

€733.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€733.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

€733.00


Check Cart for Availability

Request a Bulk Order

form

essentially salt-free, lyophilized powder

mol wt

~7000

technique(s)

affinity chromatography: suitable

color

white

UniProt accession no.

storage temp.

2-8°C

Gene Information

Amino Acid Sequence

Met-Ala-Ile-Pro-Pro-Lys-Lys-Asn-Gln-Asp-Lys

Application


  • Bioactive Peptide in Dairy Research: Investigation into the effects of bovine caseinoglycopeptide on CD4+ T cells showcased its potential to modulate immune responses, highlighting its application in nutrition and functional food development (Otani et al., 1996).

Biochem/physiol Actions

The soluble carboxyl-terminal moiety of κ-casein. This antithrombotic peptide binds to glycoprotein GPIb in the platelet membrane, thus inhibiting platelet aggregation by blocking the binding of von Willebrand factor.

Preparation Note

Prepared by a modification of the method of Alais and Jolles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Comparative study of the caseinoglycopeptides formed by the action of rennin on caseins from the cow, sheep and goat. II. Study of the non-peptide portion].
C ALAIS et al.
Biochimica et biophysica acta, 51, 315-322 (1961-08-05)
Franz-Georg Hanisch
Analytical chemistry, 83(12), 4829-4837 (2011-04-30)
The sites of mucin-type O-glycosylation are largely unpredictable, making structural analysis by mass spectrometry (MS) indispensible. On the peptide level, a site localization and characterization of O-linked glycans in situ using tandem MS with electron-transfer dissociation or matrix-assisted laser desorption
K Benschop et al.
Journal of clinical microbiology, 46(12), 3965-3970 (2008-10-24)
Human parechoviruses (HPeV) are widespread pathogens belonging to the Picornavirus family. Six genotypes, which have predominantly been isolated from children, are known. Data on prevalence of HPeV genotypes are generally based on cell culture, which may underestimate the prevalence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service