Skip to Content
Merck

Key Documents

Skip To

C7157

Coproporphyrin III tetramethyl ester

powder, ≥85% (HPLC)

Synonym(s):

Tetramethyl 3,8,13,17-tetramethyl-21H,23H-porphine-2,7,12,18-tetrapropionate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 106,00
5 mg
Check Cart for Availability
€ 358,00

About This Item

Empirical Formula (Hill Notation):
C40H46N4O8
CAS Number:
Molecular Weight:
710.82
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
EC Number:
226-869-8
MDL number:
eCl@ss:
32160703

€ 106,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Coproporphyrin III tetramethyl ester, ≥85% (HPLC)

biological source

synthetic (organic)

Quality Segment

assay

≥85% (HPLC)

form

powder

mp

168-170 °C (lit.)

solubility

chloroform: 1 mg/mL, clear

storage temp.

−20°C

SMILES string

COC(=O)CCc1c(C)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C)c5CCC(=O)OC)c(CCC(=O)OC)c4C)c(C)c3CCC(=O)OC

InChI

1S/C40H46N4O8/c1-21-25(9-13-37(45)49-5)33-18-31-23(3)26(10-14-38(46)50-6)34(43-31)19-32-24(4)28(12-16-40(48)52-8)36(44-32)20-35-27(11-15-39(47)51-7)22(2)30(42-35)17-29(21)41-33/h17-20,41,44H,9-16H2,1-8H3/b29-17-,30-17-,31-18-,32-19-,33-18-,34-19-,35-20-,36-20-

InChI key

LBHYIBKOOKXTGC-LHPWBCDESA-N

General description

One of the main tetrapyrrole compounds secreted by Rhodobacter sphaeroides in culture.

Preparation Note

Enzymatically prepared

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C4529AABH97CD7A75ATEH9C542866
description

≥85% (HPLC)

description

≥90% (HPLC)

description

-

description

-

biological source

synthetic (organic)

biological source

-

biological source

-

biological source

-

assay

≥85% (HPLC)

assay

≥90% (HPLC)

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

form

powder

form

powder

form

-

form

-

storage temp.

−20°C

storage temp.

room temp

storage temp.

-

storage temp.

-

solubility

chloroform: 1 mg/mL, clear

solubility

chloroform: 10 mg/mL, dark red

solubility

-

solubility

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters