Skip to Content
Merck

Skip To

C6269

Chromosorb® LC-9

10 μm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 752,00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
SB.52
MDL number:

€ 752,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

surface area

>350 m2/g

particle size

10 μm

density

0.48 g/cm3 at 25 °C (lit.)

Application

Separation of carbohydrates, high fructose syrup, sugars, amines, nucleotides, strongly polar compounds, carboxylic acids, polyhydroxy compounds, etc.

Preparation Note

Prepared by chemically bonding an amino type functional group on the microparticle silica substrate.

Other Notes

A polar-bonded phase on Chromosorb LC-6.

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6267LCPAK0001LCPAK00A1
particle size

10 μm

particle size

10 μm

particle size

-

particle size

-

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

300

form

solid

form

solid

form

-

form

-

surface area

>350 m2/g

surface area

>250 m2/g

surface area

-

surface area

-

density

0.48 g/cm3 at 25 °C (lit.)

density

0.45 g/cm3 (lit.)

density

-

density

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters