Skip to Content
Merck
All Photos(1)

Documents

C6171

Sigma-Aldrich

Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg

≥90% (HPLC)

Synonym(s):

Cys-Laminin A chain 2091-2108

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C82H149N31O26S
CAS Number:
Molecular Weight:
2017.32
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Quality Level

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CO)NC(=O)[C@@H](N)CS)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C82H149N31O26S/c1-12-39(6)60(78(137)105-47(21-14-16-28-84)71(130)112-58(37(2)3)76(135)100-44(11)65(124)111-59(38(4)5)77(136)110-53(33-114)73(132)99-43(10)63(122)107-52(32-57(118)119)72(131)106-51(79(138)139)24-19-31-95-82(91)92)113-75(134)55(35-116)108-64(123)41(8)96-61(120)40(7)97-68(127)50(25-26-56(86)117)104-69(128)46(20-13-15-27-83)102-70(129)49(23-18-30-94-81(89)90)101-62(121)42(9)98-67(126)48(22-17-29-93-80(87)88)103-74(133)54(34-115)109-66(125)45(85)36-140/h37-55,58-60,114-116,140H,12-36,83-85H2,1-11H3,(H2,86,117)(H,96,120)(H,97,127)(H,98,126)(H,99,132)(H,100,135)(H,101,121)(H,102,129)(H,103,133)(H,104,128)(H,105,137)(H,106,131)(H,107,122)(H,108,123)(H,109,125)(H,110,136)(H,111,124)(H,112,130)(H,113,134)(H,118,119)(H,138,139)(H4,87,88,93)(H4,89,90,94)(H4,91,92,95)/t39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,58-,59-,60-/m0/s1

InChI key

NAPSKHHWYAWPQX-YJTOHMMESA-N

Gene Information

human ... LAMA1(284217)

Amino Acid Sequence

Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg

Application

Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg has been used to culture human adenoid cystic carcinoma cells. It has also been used in the preparation of peptide-functionalized supported phospholipid bilayers.

Biochem/physiol Actions

Supports neurite outgrowth and stimulates neuronal-like process formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahu Arslan Yildiz et al.
Analytical biochemistry, 423(1), 39-45 (2012-02-07)
The analysis of membrane proteins is notoriously difficult because isolation and detergent-mediated reconstitution often results in compromising the protein structure and function. We introduce a novel strategy of combining a cell-free expression method for synthesis of a protein species coping
Vanessa Morais Freitas et al.
Virchows Archiv : an international journal of pathology, 441(6), 569-576 (2002-12-04)
We have previously demonstrated that laminin modulates the expression of adhesion molecules in an adenoid cystic carcinoma cell line (CAC2 cells). We are currently studying whether laminin can induce modifications in the overall morphology of CAC2 cells. These cells were
D Thid et al.
Journal of biomedical materials research. Part A, 84(4), 940-953 (2007-07-25)
Supported phospholipid bilayers constitute a biomimetic platform for cell behavior studies and a new approach to the design of cell culture substrates. Phosphocholine bilayers are resistant to cell attachment, but can be functionalized with bioactive molecules to promote specific cell
G C Sephel et al.
Biochemical and biophysical research communications, 162(2), 821-829 (1989-07-31)
Neurons from peripheral and central nervous tissue as well as from established cell lines respond to low concentrations of laminin with rapid extension of axon-like processes. Two sites on laminin have been identified which stimulate neurite outgrowth, the major site
Ahu Arslan Yildiz et al.
Analytical biochemistry, 423(1), 39-45 (2012-02-07)
The analysis of membrane proteins is notoriously difficult because isolation and detergent-mediated reconstitution often results in compromising the protein structure and function. We introduce a novel strategy of combining a cell-free expression method for synthesis of a protein species coping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service