Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
51111800
MDL number:
Conjugate:
unconjugated
Clone:
53103.111, monoclonal
Application:
FACS, IHC
Citations:
4
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
53103.111, monoclonal
form
lyophilized powder
species reactivity
human
technique(s)
flow cytometry: suitable using human PBMCs (1 μg/106 cells), immunohistochemistry: 25 μg/mL using frozen human tissue sections
isotype
IgG2b
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... CCR6(1235)
mouse ... Ccr6(12458)
General description
Chemokine receptor CCR6 plays an important role in adaptive immunity, recruitment of CD4+ T cells and dendritic cells to sites of epithelial inflammation. It is expressed on leukocyte subsets of T cells, B cells, immature dendritic cells, neutrophils, monocytes and NKT cells. CCR6 binds the chemokine, CCL20 (macrophage inflammatory protein-3α) and induces proinflammatory signals such as TNF-α, IFN-γ and toll-like receptor agonists that in turn activate ERK1/2 and p38 MAPK pathways. CCR6 has been linked to autoimmune diseases of multiple sclerosis, rheumatoid arthritis, psoriasis and atherosclerosis. Monoclonal Anti-CCR-6 reacts with CCR-6 transfected cells and not with the parent cell line. The antibody shows no cross-reactivity with CCR-1, CCR-2, CCR-3, CCR-4, CCR-5, CCR-7, CXCR-1, CXCR-2, CXCR-3, CXCR-4 and STRL33 transfected cells.
Immunogen
human CCR-6 transfected NSO mouse myeloma cells.
Application
Monoclonal Anti-CCR-6 antibody may be used for flow cytometry at a working concentration of 1μg/106 cells using human peripheral blood mononuclear cells. Antibody concentration of 25 μg/ml may be used for immunohistochemistry of frozen human tissue sections.
Biochem/physiol Actions
The antibody reacts with human CCR-6 transfectant cells and not the parent cell line. The antibody shows no cross-reactivity with CCR-1, CCR-2, CCR-3, CCR-4, CCR-5, CCR-7, CXCR-1, CXCR-2, CXCR-3, CXCR-4 or STRL33.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.
Preparation Note
Purified using protein G.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.