Skip to Content
Merck

Skip To

C5515

18-Crown-6

Synonym(s):

1,4,7,10,13,16-Hexaoxacyclooctadecane

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C12H24O6
CAS Number:
Molecular Weight:
264.32
EC Number:
241-473-5
UNSPSC Code:
12350000
MDL number:
Beilstein/REAXYS Number:
1619616


mp

42-45 °C (lit.)

storage temp.

2-8°C

SMILES string

O1CCOCCOCCOCCOCCOCC1

InChI

1S/C12H24O6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2

InChI key

XEZNGIUYQVAUSS-UHFFFAOYSA-N

Application

Can be useful as phase-transfer catalysts.

Other Notes

Macrocyclic polyethers with repeating (-CH2CH2O) units. The compounds are ionophoric (form stable complexes with cations).


Still not finding the right product?

Explore all of our products under 18-Crown-6


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library