Skip to Content
Merck
All Photos(1)

Key Documents

C5286

Sigma-Aldrich

(−)-β-Citronellol

Synonym(s):

β-Rhodinol, (S)-(−)-β-Citronellol, (S)-3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1721505
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

bp

221-224 °C

density

0.854 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1

InChI key

QMVPMAAFGQKVCJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renan G Brito et al.
Basic & clinical pharmacology & toxicology, 112(4), 215-221 (2012-10-06)
Citronellol (CT) is a monoterpenoid alcohol present in the essential oil of many medicinal plants, such as Cymbopogon citratus. We evaluated the antinociceptive effects of CT on orofacial nociception in mice and investigated the central pathway involved in the effect.
Paola Gonzalez-Audino et al.
Archives of dermatological research, 303(5), 361-366 (2010-12-22)
The use of botanical compounds such as essential oils has recently become the subject of great interest as a natural means of pest control because of their ovicidal, larvicidal, or adulticidal activity against various insect species including head lice. We
Karin Förster-Fromme et al.
Applied microbiology and biotechnology, 87(3), 859-869 (2010-05-22)
Terpenes are a huge group of natural compounds characterised by their predominantly pleasant smell. They are built up by isoprene units in cyclic or acyclic form and can be functionalised by carbonyl, hydroxyl or carboxyl groups and by presence of
M Spiller et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 120(7), 1461-1471 (2010-01-20)
The scent of flowers is a very important trait in ornamental roses in terms of both quantity and quality. In cut roses, scented varieties are a rare exception. Although metabolic profiling has identified more than 500 scent volatiles from rose
Yu-Wen Su et al.
Planta medica, 76(15), 1666-1671 (2010-05-28)
Geranium oil has been used traditionally for diarrhea, dermatitis, and intestinal inflammation in East Asia. The aim of this study was to determine the effects of geranium oil's characteristic components, citronellol and geraniol, on lipopolysaccharide (LPS)-induced nitric oxide (NO) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service