Skip to Content
Merck

Skip To

C4945

Sodium cacodylate trihydrate

BioXtra, ≥98%

Synonym(s):

Cacodylic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Dimethylarsonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2AsO2Na · 3H2O
CAS Number:
Molecular Weight:
214.03
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
204-708-2
Beilstein/REAXYS Number:
3702348
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Segment

assay

≥98%

form

powder

impurities

≤0.005% Phosphorus (P), ≤0.1% Insoluble matter

pH

8.0-9.0

solubility

H2O: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%, sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%, Ca: ≤0.001%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.05%, Mg: ≤0.0005%, NH4+: ≤0.05%, Pb: ≤0.001%, Zn: ≤0.0005%

application(s)

ADME/TOX

storage temp.

room temp

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].C[As](C)([O-])=O

InChI

1S/C2H7AsO2.Na.3H2O/c1-3(2,4)5;;;;/h1-2H3,(H,4,5);;3*1H2/q;+1;;;/p-1

InChI key

RLGWPHBPRCROJO-UHFFFAOYSA-M

Application

Sodium cacodylate trihydrate has been used as a buffer to fix cells for transmission electron microscopy and as a buffer to fix cells for scanning electron microscopy.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S7670C025071188
assay

≥98%

assay

≥99.0%

assay

≥98%

assay

≥99.5% (NT)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

product line

BioXtra

product line

BioXtra

product line

-

product line

BioUltra

application(s)

ADME/TOX

application(s)

-

application(s)

ADME/TOX

application(s)

-

form

powder

form

powder

form

powder

form

powder or crystals

solubility

H2O: 0.5 M, clear, colorless

solubility

H2O: 1 M, clear, colorless

solubility

water: 100 mg/mL, clear, colorless

solubility

H2O: 3 M at 20 °C, clear, colorless


Still not finding the right product?

Explore all of our products under Sodium cacodylate trihydrate


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

6131-99-3

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters