Skip to Content
Merck
All Photos(1)

Documents

C4618

Sigma-Aldrich

Monoclonal Anti-Cathepsin L antibody produced in mouse

clone CPL33/1, purified from hybridoma cell culture

Synonym(s):

Anti-CATL, Anti-CTSL, Anti-MEP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CPL33/1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~25 kDa (human cathepsin L)
antigen ~42 kDa (human pro cathepsin L)

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 0.1-0.2 μg/mL using total cell extracts of A549 cells

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTSL1(1514)

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Cathepsin L antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma CPL33/1 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with human procathepsin L.
Monoclonal Anti-Cathepsin L (mouse IgG1 isotype) is derived from the hybridoma CPL33/1 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from BALB/c mice immunized with human procathepsin L.

Specificity

The antibody recognizes the native and denaturated forms of the protein and does not cross react with human cathepsin V. Monoclonal Anti-Cathepsin L specifically recognizes human cathepsin L (∼ 25 kDa) and procathepsin L (∼ 42 kDa). Anti-Cathepsin L antibody epitope resides within amino acids of human cathepsin L (FYKE).

Immunogen

human procathepsin L. The antibody epitope resides within amino acids 258-261 of human cathepsin L (FYKE).

Application

Anti-Cathepsin L antibody has been used for immunoblotting.
Monoclonal Anti-Cathepsin L antibody produced in mouse is suitable for:
  • immunohistochemistry
  • indirect ELISA
  • western blot : 0.1-0.2 μg/mL using total cell extracts of A549 cells

Biochem/physiol Actions

Cathepsins are lysosomal proteases that play an important role in the intracellular degradation of exogenous and endogenous proteins, activation of enzyme precursors, and tumor invasion and metastasis.
Inhibition of the enzyme or the proenzyme by low molecular weight inhibitors or by specific antibodies led to a suppression of the invasive capabilities of malignant cells, or a decline in their ability to form tumors in experimental in vivo and in vitro models.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheuk-Yiu Law et al.
Biochemistry and biophysics reports, 5, 335-345 (2016-01-14)
Patients with Danon disease may suffer from severe cardiomyopathy, skeletal muscle dysfunction as well as varying degrees of mental retardation, in which the primary deficiency of lysosomal membrane-associated protein-2 (LAMP2) is considerably associated. Owing to the scarcity of human neurons
E Weber et al.
Hybridoma, 16(2), 159-166 (1997-04-01)
Mouse monoclonal antibodies directed against cathepsin L and procathepsin L have been generated. Mice were immunized with human procathepsin L purified from the cell culture medium of human nonsmall cell lung cancer cell line EPLC 32 M1. More than 400
Cysteine cathepsins and the cutting edge of cancer invasion.
Gocheva V and Joyce J A
Cell Cycle, 6(1), 60-64 (2007)
S A Igdoura et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 43(5), 545-557 (1995-05-01)
Cathepsins are specific proteases in lysosomes that participate in the degradation of proteins, some of which are derived from endocytosis. In this study we examined the immunocytochemical localization of cathepsin B and D antibodies in cells of rat testis and
Boris Turk et al.
FEBS letters, 581(15), 2761-2767 (2007-06-05)
Proteases were, for a long time, mainly considered as protein degrading enzymes. However, in the last decade this view has changed dramatically, and the focus is now on proteases as signalling molecules. One of the best examples is apoptosis, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service