Skip to Content
Merck
All Photos(1)

Documents

C4616

Sigma-Aldrich

Anti-Calcium Channel CaV3.3 (α1I) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Calcium channel CaV3.3 (α1I) is a low-voltage-activated T-type calcium channel. CaV3.3 (α1I) transcripts are detected in central nervous system as well as thyroid and adrenal glands.

Immunogen

peptide corresponding to amino acid residues 1053-1067 of rat CaV3.3. Homology with other species: human is 14/15 residues identical.

Application

Anti-Calcium Channel CaV3.3 (a1I) antibody produced in rabbit is suitable for western blot at a dilution of 1:200 using rat brain membranes.

Biochem/physiol Actions

Calcium channel CaV3.3 (α1I) is involved in neuronal excitability and is particularly recruited during the depolarizing after-potential. It is responsible for generating sustained electrical activity. Sperm specific proteins CatSper1 and CatSper2 interact with Cav3.3 and suppress Cav3.3-evoked T-type calcium channel activity.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Perez-Reyes
Physiological reviews, 83(1), 117-161 (2002-12-31)
T-type Ca2+ channels were originally called low-voltage-activated (LVA) channels because they can be activated by small depolarizations of the plasma membrane. In many neurons Ca2+ influx through LVA channels triggers low-threshold spikes, which in turn triggers a burst of action
Di Zhang et al.
The Journal of biological chemistry, 281(31), 22332-22341 (2006-06-03)
Sperm-specific CatSper1 and CatSper2 proteins are critical to sperm-hyperactivated motility and male fertility. Although architecturally resembling voltage-gated ion channels, neither CatSper1 nor CatSper2 alone forms functional ion channels in heterologous expression systems, which may be related to the absence of
Jean Chemin et al.
The Journal of physiology, 540(Pt 1), 3-14 (2002-04-03)
In several types of neurons, firing is an intrinsic property produced by specific classes of ion channels. Low-voltage-activated T-type calcium channels (T-channels), which activate with small membrane depolarizations, can generate burst firing and pacemaker activity. Here we have investigated the
Multiple Calcium Channel Types with Unique Expression Patterns Mediate Retinal Signaling at Bipolar Cell Ribbon Synapses.
Zhang, et al.
The Journal of Neuroscience, 42, 6487-6505 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service