Skip to Content
Merck
All Photos(1)

Key Documents

C4519

Sigma-Aldrich

Ceruloplasmin human

lyophilized powder, oxidase 20-75 units/mg protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 UNITS
€766.00

€766.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 UNITS
€766.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€766.00


Estimated to ship on24 May 2025


Request a Bulk Order

form

lyophilized powder

Quality Level

oxidase activity

20-75 units/mg protein

mol wt

~135 kDa

composition

Protein, ~30%

copper content

1.0-2.5 μg/mg protein

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CP(1356)

Biochem/physiol Actions

Ceruloplasmin (Cp), the principal carrier of copper in plasma, is thought to be involved in oxidative modification of lipoproteins and participate in the acute phase reaction to stress. Cp is an effective antioxidant for a variety of radicals and has a potent peroxidase activity to decompose hydrogen peroxide in the presence of reduced glutathione. It inhibits the peroxidase activity of myeloperoxidase in a concentration-dependent manner and shows selective binding to myeloperoxidase in vitro. This suggests that ceruloplasmin may participate in the clearance and inactivation of myeloperoxidase in vivo. At physiological (micromolar) concentrations it was found to inhibit relaxation of rabbit aorta induced by endothelium-dependent agonists like acetylcholine or ADP. However, it was found ineffective toward vasodilation due to direct stimulation of smooth muscle cells by nitroglycerin.
Ceruloplasmin (Cp), the principal carrier of copper in plasma, is thought to be involved in oxidative modification of lipoproteins and participate in the acute phase reaction to stress. Cp is an effective antioxidant for a variety of radicals and has a potent peroxidase activity to decompose hydrogen peroxide in the presence of reduced glutathione. It inhibits the peroxidase activity of myeloperoxidase in a concentration-dependent manner and shows selective binding to myeloperoxidase in vitro. This suggests that ceruloplasmin may participate in the clearance and inactivation of myeloperoxidase in vivo. At physiological (micromolar) concentrations it was found to inhibit relaxation of rabbit aorta induced by endothelium-dependent agonists like acetylcholine or ADP. However, it was found ineffective toward vasodilation due to direct stimulation of smooth muscle cells by nitroglycerin. Cp plays an essential role in iron homeostasis, regulating the efflux of iron from macrophages and the oxidation of Fe2+ to Fe3+. Cp deficiency leads to neurodegenerative disorders, possibly by starving neurons of iron.

Unit Definition

One unit will cause a ΔA550 of 0.01 per min using N,N-dimethyl-p-phenylenediamine as substrate at pH 5.5 and 37 °C, in a 7 mL reaction volume.

Physical form

Pale blue powder containing sodium chloride and sodium acetate

Analysis Note

Protein determined by biuret

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong Soo Joo et al.
Biological & pharmaceutical bulletin, 33(11), 1854-1860 (2010-11-05)
In pancreatic islets, free radical formation produced upon exposure to proinflammatory cytokines mediates β cell destruction, which ultimately leads to type 1 diabetes (T1D). In this study, we examined whether laccase, a family of the blue copper protein, can be
Kamil Strzelak et al.
Talanta, 214, 120881-120881 (2020-04-13)
Two analytical strategies for determination of both biocatalytic activity and concentration of ceruloplasmin conditions have been proposed. For this purpose, two constructions of fully-mechanized Multicommutated Flow Analysis (MCFA) systems were designed. The versatility of solenoid pumps and valves arrangement enabled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service