Skip to Content
Merck
All Photos(1)

Key Documents

C4021

Sigma-Aldrich

CM-Cellulose

microgranular, 25-60 μm

Synonym(s):

Carboxymethyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511

form

microgranular

particle size

25-60 μm

capacity

1.0 meq/g

InChI

1S/C6H12O6.C2H4O2/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4/h1,3-6,8-12H,2H2;1H3,(H,3,4)

InChI key

VJHCJDRQFCCTHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

CM Cellulose products are used in protein chromatography, ion exchange chromatography and cation exchange media. CM Cellulose has been used in studies to inform the suppressed proliferation of hepatoma and leukemic cells and the inhibition of the human immunodeficiency virus type 1 (HIV-1).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingying Wu et al.
Carbohydrate research, 346(16), 2576-2581 (2011-10-04)
The isolation of a dimeric 29.8-kDa lectin (LFL) from dried Lactarius flavidulus fruit bodies is reported herein. The chromatographic procedure utilized comprised anion-exchange chromatography on DEAE-cellulose, cation-exchange chromatography on CM-cellulose, anion-exchange chromatography on SP-Sepharose, and gel filtration by fast protein
Raíssa Debacker Moura et al.
Journal of microbiological methods, 173, 105915-105915 (2020-04-08)
Fusarium guttiforme and Fusarium ananatum are the etiological agents of fusariosis and fruitlet core rot in pineapple, respectively, producing mycotoxins that are harmful to the health of consumers. These two fungi are morphologically similar and difficulty in obtaining macroconidia of
Yingying Wu et al.
The Journal of antibiotics, 65(2), 67-72 (2011-12-22)
A 14.6-kDa RNase, with a pH optimum of 5.5 and a temperature optimum of 70 °C, was isolated from dried fruiting bodies of the edible mushroom Lactarius flavidulus. The purification procedure involved, in succession, ion exchange chromatography on DEAE-cellulose, CM-cellulose and
T Subramani et al.
The protein journal, 31(2), 120-128 (2011-12-17)
Carboxylesterases are hydrolases which catalyze the hydrolysis of various types of esters. Carboxylesterase from the seeds of Jatropha curcas has been purified to homogeneity using ammonium sulfate fractionation, CM-cellulose chromatography, Sephadex G-100 chromatography and preparative polyacrylamide gel electrophoresis (PAGE). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service