Skip to Content
Merck
All Photos(1)

Key Documents

C-39175

Sigma-Aldrich

Small Airway Epithelial Cell Growth Medium

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-21270

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€181.00

€181.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
€181.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

€181.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Small Airway Epithelial Cell Growth Medium SupplementMix (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-21270

Application

Small Airway Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the distal respiratory system. The optimal adjustment of growth factors in this medium ensures excellent in vitro growth. The medium is optimized for primary human cells but can also be used for porcine small airway epithelial cells.Recommended for:Human Small Airway Epithelial Cells (HSAEpC)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Small Airway Epithelial Cell Growth Medium are subjected to comprehensive quality control tests using primary human small airway epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested small airway epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 10 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.5 µg / mlEpinephrine: 0.5 µg / mlTriiodo-L-thyronine: 6.7 ng / mlTransferrin (recombinant human): 10 µg / mlRetinoic Acid: 0.1 ng / mlBovine Serum Albumin-Fatty Acid Free (BSA-FAF): 2.5 mg/ml

Storage and Stability

Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service