Skip to Content
Merck
All Photos(1)

Key Documents

C-39011

Sigma-Aldrich

Keratinocyte Growth Medium 2

SupplementPack containing all media supplements as individual vials, 1 Pack for 500 ml

Synonym(s):

Recommended with C-20211

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81
Pricing and availability is not currently available.

form

liquid

packaging

pkg of 1 kit

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Keratinocyte Growth Medium 2 SupplementPack (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-20211

Application

Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells. The medium is optimized for primary human cells but can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.Recommended for:Normal Human Epidermal Keratinocytes (NHEK), juvenile foreskinNormal Human Epidermal Keratinocytes (NHEK), adult donor

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Keratinocyte Growth Medium 2 are subjected to comprehensive quality control tests using primary human keratinocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested keratinocytes. Approved inhouse lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 0.125 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.33 µg / mlEpinephrine: 0.39 µg / mlTransferrin (recombinant human): 10 µg / mlCaCl2: 0.06 mMNote: CaCl2 is provided as separate stock solution. This allows users to adjust the CaCl2 concentration individually. To ensure the maximal growth performance of normal human Keratinocytes, PromoCell recommends adding the entire 60 μlCaCl2 stock solution, which results in final 0.06 mM CaCl2 concentration. In case a higher CaCl2 concentration is required (e.g. differentiation experiments), a 0.5 M CaCl2-Solution can be ordered separately (C-34006).

Storage and Stability

Store the SupplementPack at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human keratinocytes and serum-free keratinocyte growth media for the feeder cell-free in vitro cultivation of epidermal keratinocytes. Protocols for handling, cell passaging, media, and product use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service