Merck
Skip To
C3858
Scienceware® cloning cylinders, polystyrene
I.D. × H 9.5 mm × 11 mm
Synonym(s):
cloning cylinder, cloning equipment, 378470300, F37847-0300, clone isolation chamber, cloning flask
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
50 ea
Check Cart for Availability
€ 135,00
About This Item
NACRES:
NB.22
UNSPSC Code:
41106618
€ 135,00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
sterility
sterile
packaging
pack of 50 ea
manufacturer/tradename
Bel-Art F37847-0300
I.D. × H
9.5 mm × 11 mm
General description
To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
C7983C3983Z370789
manufacturer/tradename
Bel-Art F37847-0300
manufacturer/tradename
Bel-Art F37847-0100
manufacturer/tradename
Bel-Art F37847-0200
manufacturer/tradename
Bel-Art H37847-0000
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
packaging
pack of 50 ea
packaging
pack of 50 ea
packaging
pack of 50 ea
packaging
box of 3 bags
I.D. × H
9.5 mm × 11 mm
I.D. × H
4.7 mm × 8 mm
I.D. × H
6.4 mm × 8 mm
I.D. × H
-
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)