Skip to Content
Merck
All Photos(1)

Documents

C3090

Sigma-Aldrich

pcDNA 1.1 (−)/Amp/CAT control vector

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

storage temp.

−20°C

Specificity

The MCS contains unique BamH I, EcoR I, EcoR V, Hind III, Not I, Nsi I, Sph I, Xba I, and Xho I restriction sites, and redundant BstX I sites.

Features and Benefits

A 4.8-kb vector for transient, high-level protein expression in mammalian cells. The vector incorporates the strong cytomegalovirus (CMV) enhancer-promoter sequence for high-level expression, SV40 and polyoma origins of replication to facilitate episomal replication, an ampicillin resistance gene for selection in E. coli, and the col E1 origin and supF suppressor tRNA for growth and maintenance in appropriate strains of E. coli.

Other Notes

This is the pcDNA1.1/Amp vector with an insert containing the chloramphenicol acetyltransferase gene.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service