C30200-HH
DMT-dC(tac)-CPG
pore size 500 Å, 30-40μmol/g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116105
PubChem Substance ID:
type
for DNA synthesis
product line
Proligo Reagents
Assay
≥95.0% (Nucleoside Purity, reversed phase HPLC)
packaging
pkg of 1 × 1 g C302001-01
pkg of 1 × 10 g C302010-01
pore size
500 Å
capacity
≥99.0 % (Coupling Efficiency)
λ
conforms (UV/VIS Identity)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service