Skip to Content
Merck

Skip To

C2911

Cobalt(II) chloride hexahydrate

suitable for plant cell culture

Synonym(s):

Cobaltous chloride hexahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Check Cart for Availability
€ 113,00

About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
NACRES:
NA.71
PubChem Substance ID:
UNSPSC Code:
12352207
EC Number:
231-589-4
MDL number:
Form:
powder

€ 113,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

40 mmHg ( 0 °C)

Quality Segment

form

powder

technique(s)

cell culture | plant: suitable

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Application


  • Bis(imino)-6,7-dihydro-5H-quinoline-cobalt complexes as highly active catalysts for the formation of vinyl-terminated PE waxes.: Research demonstrates the application of Cobalt(II) chloride hexahydrate in synthesizing novel cobalt complexes, enhancing polymerization processes and introducing new pathways in materials science (Han et al., 2021).

  • Synthesis, Characterization, and Biological Activities of Novel Vanadium(IV) and Cobalt(II) Complexes.: Highlights the role of Cobalt(II) chloride hexahydrate in the synthesis and biological evaluation of metal complexes, suggesting potential therapeutic and bioactive properties (Damena et al., 2022).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C8661202185255599
form

powder

form

powder

form

crystals or chunks

form

powder, crystals or chunks

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

vapor pressure

40 mmHg ( 0 °C)

vapor pressure

40 mmHg ( 0 °C)

vapor pressure

40 mmHg ( 0 °C)

vapor pressure

40 mmHg ( 0 °C)

technique(s)

cell culture | plant: suitable

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

cell culture trace element, trace elements

cell culture trace element, trace elements

-

-


Still not finding the right product?

Explore all of our products under Cobalt(II) chloride hexahydrate


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7791-13-1

CAS No.

7791-13-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters