Skip to Content
Merck
All Photos(2)

Key Documents

C2851

Sigma-Aldrich

Anti-CaM Kinase IV (CaMKIV) (AY-18) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
€446.00

€446.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
0.2 ML
€446.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

€446.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 65 kDa

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10,000 using sections of rat cerebellum
microarray: suitable
western blot: 1:20,000-1:1,000 using rat brain extract and Jurkat, whole cell extract, respectively

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK4(814)
mouse ... Camk4(12326)
rat ... Camk4(25050)

General description

CaM Kinase IV is a Ca2+/calmodulin-dependent kinase that belongs to serine/threonine protein kinases family. CaM Kinase IV is expressed in the brain and thymus and is localized in cellular cytoplasm and nucleus. It facilitates the phosphorylation of several substrates in brain like-synapsin I, microtubule associated protein (MAP2), tau and tyrosine hydroxylase. Anti-caM kinase IV (CaMKIV) (AY-18) antibody can be used in western blotting (diluted 1:1000 and 1: 20,000) using human T-cell leukemia Jurkat whole cell extract and rat brain extract. It can also be used in microarray. Rabbit anti-caM kinase IV (CaMKIV) (AY-18) antibody reacts specifically with CaMKIV of rat but cross reacts with human CaMKIV.

Immunogen

Peptide corresponding to amino acids 457-474 of rat CaM kinase IV conjugated to KLH. This sequence has extensive homology to mouse (80%) and human (70%) CaMKIV.

Application

Anti-caM kinase IV (CaMKIV) (AY-18) antibody (diluted 1:500) can be used as a primary antibody in immunohistochemical studies. It can also be used in immunoblotting to detect and localize CAMKIV (65 kDa).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M V Frangakis et al.
The Journal of biological chemistry, 266(26), 17592-17596 (1991-09-15)
The regional and tissue-specific expression of the Ca2+/calmodulin-dependent protein kinase, CaM kinase-Gr, were examined. The Mr 65,000 alpha-polypeptide of CaM kinase-Gr is expressed ubiquitously in different anatomical regions of rat brain, whereas an additional Mr 67,000 beta-polypeptide is observed solely
Y Nakamura et al.
Neuroscience, 68(1), 181-194 (1995-09-01)
We observed the distribution pattern of Ca2+/calmodulin-dependent protein kinase IV in rat brain and spinal cord using an immunohistochemical method by light and electron microscopy. Particularly strong immunoreactivity was detected in the telencephalic structures such as the olfactory bulb, cerebral
Nicholas F Trojanowski et al.
eNeuro, 8(4) (2021-05-19)
Pyramidal neurons in rodent visual cortex homeostatically maintain their firing rates in vivo within a target range. In young cultured rat cortical neurons, Ca2+/calmodulin-dependent kinase IV (CaMKIV) signaling jointly regulates excitatory synaptic strength and intrinsic excitability to allow neurons to
Hong-Zhen Hu et al.
The Journal of pharmacology and experimental therapeutics, 309(1), 310-319 (2004-01-14)
Intracellular recording methods with "sharp" microelectrodes were used to study signal transduction mechanisms underlying the excitatory action of bradykinin (BK) in morphologically identified neurons in the small intestinal submucosal plexus. Exposure to BK evoked slowly activating membrane depolarization and enhanced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service