Skip to Content
Merck

Skip To

C-27015

Osteoblast Growth Medium

Basal Medium, phenol red-free, 500 ml

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Check Cart for Availability
€ 265,00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207

€ 265,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Osteoblast Basal Medium, phenol red-free (supplements sold separately)

Application

Osteoblast Growth Medium has been optimized for the in vitro cultivation of human osteoblasts. The fetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimized for primary human cells, but can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.Recommended for:Human Osteoblasts (HOB)

Preparation Note

Click here for more information.
Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C

Analysis Note

All lots of PromoCell Osteoblast Growth Medium are subjected to comprehensive quality control tests using primary human osteoblasts. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested osteoblasts. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Other Notes

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixSupplementMix: Contains all media supplements premixed in one vial


Still not finding the right product?

Explore all of our products under Osteoblast Growth Medium


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters